Learn More
Sino Biological Recombinant Human HSP70 Protein (His Tag)
Supplier:  Sino Biological 11660H07B100
Encompass_Preferred
A DNA sequence encoding the human HSPA1A (NP_005337.2) (Ala2-Asp641) was expressed, with a polyhistidine tag at the N-terminus. HSPA1A (NP_005336.3) and HSPA1B (NP_005337.2) sequences are identical.
Catalog No. 50-161-7113