Learn More
Sino Biological Recombinant Human HSP70 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11660H07H50
Encompass_Preferred
A DNA sequence encoding the human HSPA1A (NP_005336.3) (Ala2-Asp641) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7116