Learn More
Sino Biological Recombinant Human LOXL2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11664H08S50
Encompass_Preferred
A DNA sequence encoding the human LOXL2 (Q9Y4K0) (Met1-Gln774) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7126