Learn More
Sino Biological Recombinant Human Protein Kinase D2/PRKD2 Protein (His & GST Tag)
Supplier:  Sino Biological 11671H20B50
Encompass_Preferred
A DNA sequence encoding the amino acid sequence (Met 1-Leu 878) of human PRKD2 (NP_057541.2) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7130