Learn More
Sino Biological Recombinant Human IL-12 R beta 1/IL12RB1 Protein (ECD, His Tag)
Supplier:  Sino Biological 11674H08H100
Encompass_Preferred
A DNA sequence encoding the human IL12RB1 isoform 1 (NP_005526.1) extracellular domain (Met1-Glu540) was expressed fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7131