Learn More
Sino Biological HIV-1 gp140 Protein (group M, subtype CRF07_BC) (Fc Tag)
Supplier:  Sino Biological 11677V02H100
Encompass_Preferred
A DNA sequence encoding the HIV-1 envelope glycoprotein gp160 extracellular domain (Ala 29-Ser 661), termed as gp140, was fused with the Fc region of human IgG1 at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-7140