Learn More
Sino Biological HIV-1 gp140 Protein (group M, subtype CRF07_BC) (His Tag)
Supplier:  Sino Biological 11677V08H100
Encompass_Preferred
A DNA sequence encoding the HIV-1 envelope glycoprotein gp160 extracellular domain (Val30-Tyr677), termed as gp140, was fused with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7144