Learn More
Sino Biological Recombinant Human USP7 Protein (aa 208-560, His & GST Tag)
Supplier:  Sino Biological 11681H20B100
Encompass_Preferred
A DNA sequence encoding the amino acid sequence (Lys 208-Glu 560) of human USP7 (NP_003461.2) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7154