Learn More
Sino Biological Recombinant Human RANKL/TNFSF11 Protein (mFc Tag)
Supplier:  Sino Biological 11682H04H100
Encompass_Preferred
A DNA sequence encoding the human TNFSF11 (AAC51762.1) (Gly63-Asp244) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-7163