Learn More
Sino Biological Influenza A H1N1 (A/New Caledonia/20/1999) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11683V08H1100
Encompass_Preferred
A DNA sequence encoding the influenza hemagglutinin (A/New Caledonia/20/1999 (H1N1)) (Met 1-Arg 343) (AAP34324.1), (termed as HA1) was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7177