Learn More
Sino Biological Influenza A H1N1 (A/Puerto Rico/8/1934) Hemagglutinin / HA Protein (ECD,His Tag)
Supplier:  Sino Biological 11684V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Puerto Rico/8/1934 (H1N1)) hemagglutinin (ABD77675.1) (Met1-Gln528) was expressed fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7183