Learn More
Sino Biological Influenza A H1N3 (A/duck/NZL/160/1976) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11685V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/duck/NZL/160/1976 (H1N3)) hemagglutinin (ABB20429.1) (Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7188