Learn More
Sino Biological Influenza A H2N2 (A/Canada/720/2005) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11688V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Canada/720/2005(H2N2)) hemagglutinin (AAY28987.1) (Met1-Gln525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7202