Learn More
Sino Biological Influenza A H2N2 (A/Canada/720/2005) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11688V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 340) of the influenza A hemagglutinin (A/Canada/720/2005 (H2N2)) (AAY28987.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7207