Learn More
Sino Biological Influenza A H5N1 (A/Hong Kong/483/1997) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11689V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 531) of the influenza A H5N1 hemagglutinin (A/Hong Kong/483/1997 (H5N1)) (AAC32099.1), (HA1+HA2, uncleaved, cleavage site mutated (RRRKKR-TETR) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7211