Learn More
Sino Biological Influenza A H5N1 (A/Hong Kong/483/1997) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11689V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 339) of the influenza A hemagglutinin (A/Hong Kong/483/1997 (H5N1)) (AAC32099.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7212