Learn More
Sino Biological Influenza A H5N1 (A/goose/Guiyang/337/2006) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11690V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 339) of the influenza A H5N1 hemagglutinin (A/goose/Guiyang/337/2006) (ABJ96698.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7216