Learn More
Sino Biological Recombinant Human SLAMF7/CRACC/CS1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11691H08H100
Encompass_Preferred
A DNA sequence encoding the human SLAMF7 (NP_067004.3) extracellular domain (Met 1-Met 226) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7219