Learn More
Sino Biological Influenza A H1N1 (A/WSN/1933) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11692V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/WSN/1933(H1N1)) hemagglutinin (ACF54598.1) (Met1-Gln528), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7221