Learn More
Sino Biological Influenza A H1N1 (A/WSN/1933) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11692V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 343) of the influenza hemagglutinin (A/WSN/1933 (H1N1)) (ACF54598.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7226