Learn More
Sino Biological Influenza A H10N3 (A/duck/Hong Kong/786/1979) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11693V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ile 525) of the influenza A H10N3 hemagglutinin (A/duck/Hong Kong/786/1979 (H10N3)) (BAF46762.1) (HA1+HA2, uncleaved) was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7232