Learn More
Sino Biological Influenza A H5N1 (A/Japanese white-eye/Hong Kong/1038/2006) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Catalog No. 501617239
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Glu 340) of the influenza A hemagglutinin (A/Japanese white-eye/Hong Kong/1038/2006 (H5N1)) (ABJ96775.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7239 Supplier Sino Biological Supplier No. 11694V08H1100
A DNA sequence encoding the N-terminal segment (Met 1-Glu 340) of the influenza A hemagglutinin (A/Japanese white-eye/Hong Kong/1038/2006 (H5N1)) (ABJ96775.1), termed as HA1, was fused with a C-terminal polyhistidine tag.