Learn More
Sino Biological HIV-1 p24 Protein (group M, subtype B, strain 92418) (His Tag)
Supplier:  Sino Biological 11695V08E100
Encompass_Preferred
A DNA sequence encoding the amino acid (Pro 133-Leu 363) of HIV-1 gag protein (ACI05538.1), known as core nucleocapsid protein P24 was expressed, with a polyhistidine tag at the C-terminus and a Met at the N-terminus.
Catalog No. 50-161-7242