Learn More
Sino Biological Influenza A H5N3 (A/duck/Hokkaido/167/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11696V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 527) of the influenza A H5N3 Hemagglutinin (A/duck/Hokkaido/167/2007) (BAG07130.2), was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7247