Learn More
Sino Biological Influenza A H5N1 (A/Egypt/2321-NAMRU3/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11697V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 531) of the influenza A H5N1 Hemagglutinin (A/Egypt/2321-NAMRU3/2007) (ABP96850.1) (HA1+HA2, uncleaved, with the cleavage site mutated (RRRKR-TETR) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7252