Learn More
Sino Biological Influenza A H5N1 (A/Egypt/2321-NAMRU3/2007) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11697V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Glu 340) of the influenza A H5N1 Hemagglutinin (A/Egypt/2321-NAMRU3/2007) (ABP96850.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7254