Learn More
Sino Biological Influenza A H5N1 (A/duck/Hunan/795/2002) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11698V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 531) of the influenza A Hemagglutinin (A/duck/Hunan/795/2002 (H5N1)) (ACA47835.1) (HA1+HA2, uncleaved,with the cleavage site mutated (RRRKKR-TETR)) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7257