Learn More
Sino Biological Influenza A H5N1 (A/duck/Hunan/795/2002) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11698V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Glu 340) of the influenza hemagglutinin (A/duck/Hunan/795/2002 (H5N1)) (ACA47835.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7258