Learn More
Sino Biological Influenza A H5N2 (A/American green-winged teal/California/HKWF609/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11699V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 527) of the influenza A hemagglutinin (A/American green-winged teal/California/HKWF609/2007(H5N2)) (ACF47563.1) (HA1+HA2, uncleaved), was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7262