Learn More
Sino Biological Influenza A H5N1 (A/Common magpie/Hong Kong/2256/2006) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11700V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 530) of the influenza hemagglutinin (A/Common magpie/Hong Kong/2256/2006 (H5N1)) (ABJ96777.1) (HA1+HA2, uncleaved,with the cleavage site mutated (RRRKR-TETR)) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7267