Learn More
Sino Biological Influenza A H1N2 (A/swine/Guangxi/13/2006) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11703V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 529) of the influenza A hemagglutinin (A/swine/Guangxi/13/2006 (H1N2)) (ABQ42444.1) (HA1+HA2, uncleaved) was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7279