Learn More
Sino Biological Influenza A H11N2 (A/duck/Yangzhou/906/2002) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11705V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/duck/Yangzhou/906/2002(H11N2)) hemagglutinin (AAY85533.1) (Met1-Lys528), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7287