Learn More
Sino Biological Influenza A H11N2 (A/duck/Yangzhou/906/2002) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11705V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Lys 528) of the influenza hemagglutinin (A/duck/Yangzhou/906/2002 (H11N2)) (AAY85533.1) (HA1+HA2, uncleaved), was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7290