Learn More
Sino Biological Influenza A H4N6 (A/Swine/Ontario/01911-1/99) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11706V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ile 528) of the influenza A hemagglutinin (A/Swine/Ontario/01911-1/99 (H4N6)) (AAG17429.1) (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7295