Learn More
Sino Biological Influenza A H3N2 (A/Aichi/2/1968) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11707V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Aichi/2/1968 (H3N2)) hemagglutinin (P03437.1) (Met1-Trp530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7300