Learn More
Sino Biological Influenza A H3N2 (A/Aichi/2/1968) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 11707V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Trp 530) of the influenza A hemagglutinin (A/Aichi/2/1968 (H3N2)) (AAA43178.1) (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7303