Learn More
Sino Biological Influenza A H1N1 (A/Solomon Islands/3/2006) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11708V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 343) of the influenza A H1N1 hemagglutinin (A/Solomon Islands/3/2006 (H1N1)) (ABU99109.1), was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7312