Learn More
Sino Biological Influenza A H5N1 (A/whooper swan/Mongolia/244/2005) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11709V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 531) of the influenza hemagglutinin (A/whooper swan/Mongolia/244/2005 (H5N1)) (ACZ36881.1), (HA1+HA2, uncleaved, cleavage site mutated (RRRRKR-TETR)) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7315