Learn More
Sino Biological Influenza A H5N1 (A/Cambodia/R0405050/2007) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 11710V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Cambodia/R0405050/2007(H5N1)) hemagglutinin (ACI06178.1, with cleavage site mutations (GRRKKR to TETR)) (Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7319