Learn More
Sino Biological Influenza A H5N1 (A/Cambodia/R0405050/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11710V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 531) of the influenza A hemagglutinin (A/Cambodia/R0405050/2007 (H5N1)) (ACI06178.1), (HA1+HA2, uncleaved, cleavage site mutated (GRRKKR-TETR)) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7321