Learn More
Sino Biological Influenza A H5N1 (A/Cambodia/R0405050/2007) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11710V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Glu 340) of the influenza A Hemagglutinin H5N1 (A/Cambodia/R0405050/2007 (H5N1)) (ACI06178.1), termed as HA1, was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7323