Learn More
Sino Biological Influenza A H16N3 (A/black-headed gull/Sweden/5/99) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11711V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/black-headed gull/Sweden/5/99 (H16N3)) hemagglutinin (AAV91217.1) (Met1-Lys529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7326