Learn More
Sino Biological Influenza A H16N3 (A/black-headed gull/Sweden/5/99) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11711V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 343) of the influenza A H16N3 hemagglutinin (A/black-headed gull/Sweden /5/99 (H16N3)) (AAV91217.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7330