Learn More
Sino Biological Influenza A H5N1 (A/chicken/India/NIV33487/06) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11712V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/chicken/India/NIV33487/06 (H5N1)) hemagglutinin (ABQ45850.1) (Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7333