Learn More
Sino Biological Influenza A H5N1 (A/chicken/India/NIV33487/06) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11712V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 531) of the influenza A hemagglutinin (A/chicken/India/NIV33487/06 (H5N1)) (ABQ45850.1) (HA1+HA2, uncleaved, cleavage site mutated (RRRKKR-TETR)) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7336