Learn More
Sino Biological Influenza A H5N1 (A/Hong kong/213/2003) Hemagglutinin / HA (S28W) Protein (ECD, His Tag)
Supplier:  Sino Biological 11713V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Hong kong/213/2003(H5N1)) hemagglutinin (ABP51975.1, with mutation Ser 28 Trp and cleavage site mutations (RRRKKR to TETR)) (Met1-Gln531), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7340