Learn More
Sino Biological Influenza A H4N6 (A/mallard/Ohio/657/2002) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11714V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 343) of the influenza A H4N6 hemagglutinin (A/mallard/Ohio/657/2002 (H4N6)) (ABI47995.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7350