Learn More
Sino Biological Influenza A H3N2 (A/Wyoming/03/2003) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11715V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Trp 530) of the influenza A H3N2 hemagglutinin (A/Wyoming/03/2003 (H3N2)) (ABX10525.1) (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7356