Learn More
Sino Biological Influenza A H3N2 (A/Wyoming/03/2003) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11715V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 345) of the influenza A H3N2 hemagglutinin (A/Wyoming/03/2003 (H3N2)) (ABX10525.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7358