Learn More
Sino Biological Influenza B (B/Malaysia/2506/2004) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11716V08H100
Encompass_Preferred
A DNA sequence encoding the the influenza hemagglutinin (B/Malaysia/2506/2004) (ACO05957.1) (Met 1-Ala 556) (HA1+HA2, uncleaved), was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7364